missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromophenyl)naphthalene, 98%, Thermo Scientific™

Catalog Number 15441479
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
667940-23-0
C16H11Br
283.17
MFCD16658911
FWPXWVYUNHYGPE-UHFFFAOYSA-N
2-3-bromophenyl naphthalene, naphthalene, 2-3-bromophenyl, 2 3-bromophenyl naphthalene, ksc279g1j
23080773
2-(3-bromophenyl)naphthalene
BrC1=CC=CC(=C1)C1=CC2=CC=CC=C2C=C1
This item is not returnable. View return policy
667940-23-0
C16H11Br
283.17
MFCD16658911
FWPXWVYUNHYGPE-UHFFFAOYSA-N
2-3-bromophenyl naphthalene, naphthalene, 2-3-bromophenyl, 2 3-bromophenyl naphthalene, ksc279g1j
23080773
2-(3-bromophenyl)naphthalene
BrC1=CC=CC(=C1)C1=CC2=CC=CC=C2C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Beilstein 11441102
Formula Weight 283.17
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-(3-Bromophenyl)naphthalene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'