missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Bromophenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64943.06

Code : NEW

Additional Details : CAS Number : 77738-96-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15492369

  • £543.78 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Bromophenyl)benzimidazole
77738-96-6
273.13
MFCD00602186

Specifications

2-(3-Bromophenyl)benzimidazole
5g
95%
MFCD00602186
77738-96-6
Solid
273.13
Videos
SDS