missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Bromophenyl)-1-phenylbenzimidazole, 98%

Brand:  Alfa Aesar™ H64595.03

Code : NEW

Additional Details : CAS Number : 760212-40-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15451869

  • £23.96 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Bromophenyl)-1-phenylbenzimidazole
349.23
14489825
760212-40-6
MFCD12911655

Specifications

2-(3-Bromophenyl)-1-phenylbenzimidazole
1g
98%
MFCD12911655
760212-40-6
Solid
349.23
14489825
Videos
SDS