missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromophenoxy)pyridine, 97%, Thermo Scientific™

Catalog Number p-3484865
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
92545-83-0
C11H8BrNO
250.10
MFCD08741430
CXEIVVAAUANDIA-UHFFFAOYSA-N
2-3-bromophenoxy pyridine, 2-3-bromophenoxyl pyridine, pyridine, 2-3-bromophenoxy, 3-bromophenyl pyridin-2-yl ether, 3-bromo-1-2-pyridyloxy benzene, 1-bromo-3-pyridin-2-yl oxy benzene
21616612
2-(3-bromophenoxy)pyridine
BrC1=CC=CC(OC2=CC=CC=N2)=C1
This item is not returnable. View return policy
92545-83-0
C11H8BrNO
250.10
MFCD08741430
CXEIVVAAUANDIA-UHFFFAOYSA-N
2-3-bromophenoxy pyridine, 2-3-bromophenoxyl pyridine, pyridine, 2-3-bromophenoxy, 3-bromophenyl pyridin-2-yl ether, 3-bromo-1-2-pyridyloxy benzene, 1-bromo-3-pyridin-2-yl oxy benzene
21616612
2-(3-bromophenoxy)pyridine
BrC1=CC=CC(OC2=CC=CC=N2)=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 250.09
Percent Purity 97%
Chemical Name or Material 2-(3-Bromophenoxy)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'