Learn More
2-(3-Bromophenoxy)-6-methylpyrazine, 97%, Maybridge
Brand: Maybridge CC62610DE
Code : 31
Additional Details : CAS Number : 915707-60-7 Weight : 0.05500kg Transport : UN number : 2811 Chem class : 1 Pack group : III
Quantity | 5g |
---|---|
Packaging | Amber Glass Bottle |
Chemical Identifiers
2-(3-Bromophenoxy)-6-methylpyrazine | |
96.5 | |
97% | |
C11H9BrN2O | |
MFCD09702405 |
915707-60-7 | |
100.0 | |
97% | |
265.11 |
Specifications
2-(3-Bromophenoxy)-6-methylpyrazine | |
96.5 | |
Amber Glass Bottle | |
97% | |
C11H9BrN2O | |
MFCD09702405 |
915707-60-7 | |
100.0 | |
5g | |
97% | |
265.11 |