missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-phenylpropionamide, 95%

Brand:  Alfa Aesar™ H65214.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15442489

  • £119.78 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-phenylpropionamide
329.39
MFCD03864483

Specifications

2-(3-Benzoylphenyl)-N-phenylpropionamide
95%
MFCD03864483
Solid
329.39
1g
Videos
SDS