missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(5-chloro-2-methylphenyl)propionamide, 95%

Brand:  Alfa Aesar™ H65908.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429996

  • £109.18 / 1g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-(5-chloro-2-methylphenyl)propionamide
377.86
MFCD28098839

Specifications

2-(3-Benzoylphenyl)-N-(5-chloro-2-methylphenyl)propionamide
95%
MFCD28098839
Solid
377.86
1g
Videos
SDS