missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(4-tert-butylbenzyl)propionamide, 95%

Brand:  Alfa Aesar™ H65042.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449336

  • £109.18 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-(4-tert-butylbenzyl)propionamide
399.52
MFCD28098741

Specifications

2-(3-Benzoylphenyl)-N-(4-tert-butylbenzyl)propionamide
95%
MFCD28098741
Solid
399.52
1g
Videos
SDS