missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(4-fluoro-2-methylphenyl)propionamide, 95%

Brand:  Alfa Aesar™ H65127.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429396

  • £119.78 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-(4-fluoro-2-methylphenyl)propionamide
361.41
MFCD28098752

Specifications

2-(3-Benzoylphenyl)-N-(4-fluoro-2-methylphenyl)propionamide
95%
MFCD28098752
Solid
361.41
1g
Videos
SDS