missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(4-ethylphenyl)propionamide, 95%

Brand:  Alfa Aesar™ H65349.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409586

  • £119.78 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-(4-ethylphenyl)propionamide
357.44
MFCD28098787

Specifications

2-(3-Benzoylphenyl)-N-(4-ethylphenyl)propionamide
95%
MFCD28098787
Solid
357.44
1g
Videos
SDS