missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-[4-chloro-2-(trifluoromethyl)phenyl]propionamide, 95%

Brand:  Alfa Aesar™ H65393.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499606

  • £91.90 / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-[4-chloro-2-(trifluoromethyl)phenyl]propionamide
431.83
MFCD05156038

Specifications

2-(3-Benzoylphenyl)-N-[4-chloro-2-(trifluoromethyl)phenyl]propionamide
95%
MFCD05156038
Solid
431.83
1g
Videos
SDS