missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(4-chloro-2-fluorophenyl)propionamide, 95%

Brand:  Alfa Aesar™ H65588.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499756

  • £99.64 / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-(4-chloro-2-fluorophenyl)propionamide
381.83
MFCD28098807

Specifications

2-(3-Benzoylphenyl)-N-(4-chloro-2-fluorophenyl)propionamide
95%
MFCD28098807
Solid
381.83
1g
Videos
SDS