missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(4-bromo-3-fluorophenyl)propionamide, 95%

Brand:  Alfa Aesar™ H65052.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15419346

  • £79.71 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-(4-bromo-3-fluorophenyl)propionamide
426.28
MFCD28098743

Specifications

2-(3-Benzoylphenyl)-N-(4-bromo-3-fluorophenyl)propionamide
95%
MFCD28098743
Solid
426.28
1g
Videos
SDS