missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-[4-bromo-2-(trifluoromethyl)phenyl]propionamide, 95%

Brand:  Alfa Aesar™ H65369.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459596

  • £121.90 / 1g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-[4-bromo-2-(trifluoromethyl)phenyl]propionamide
476.29
MFCD28098791

Specifications

2-(3-Benzoylphenyl)-N-[4-bromo-2-(trifluoromethyl)phenyl]propionamide
95%
MFCD28098791
Solid
476.29
1g
Videos
SDS