missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(2-methylphenyl)propionamide, 95%

Brand:  Alfa Aesar™ H65057.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15479346

  • £56.92 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-(2-methylphenyl)propionamide
343.32
MFCD03838502
2-(3-Benzoylphenyl)-N-(o-tolyl)propionamide

Specifications

2-(3-Benzoylphenyl)-N-(2-methylphenyl)propionamide
95%
MFCD03838502
1g
Solid
343.32
2-(3-Benzoylphenyl)-N-(o-tolyl)propionamide
Videos
SDS