missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(2-fluoro-5-methylphenyl)propionamide, 95%

Brand:  Alfa Aesar™ H65901.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449986

  • £127.20 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-(2-fluoro-5-methylphenyl)propionamide
361.41
MFCD28098838

Specifications

2-(3-Benzoylphenyl)-N-(2-fluoro-5-methylphenyl)propionamide
95%
MFCD28098838
Solid
361.41
1g
Videos
SDS