missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(2,5-dimethylphenyl)propionamide, 95%

Brand:  Alfa Aesar™ H65566.03

UNSPSC : 12352116

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409746

  • £85.44 / 1g


Estimated Shipment date
from Supplier 07-09-2017
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-(2,5-dimethylphenyl)propionamide
357.44
MFCD04151245

Specifications

2-(3-Benzoylphenyl)-N-(2,5-dimethylphenyl)propionamide
95%
MFCD04151245
Solid
357.44
1g
Documentation

Technical Documents