missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(2,3-dimethylphenyl)propionamide, 95%

Brand:  Alfa Aesar™ H65914.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15400007

  • £119.78 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-(2,3-dimethylphenyl)propionamide
357.44
MFCD04090496

Specifications

2-(3-Benzoylphenyl)-N-(2,3-dimethylphenyl)propionamide
95%
MFCD04090496
Solid
357.44
1g
Videos
SDS