missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(2,3-dichlorophenyl)propionamide, 95%

Brand:  Alfa Aesar™ H65146.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429416

  • £119.78 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-(2,3-dichlorophenyl)propionamide
398.28
MFCD05156077

Specifications

2-(3-Benzoylphenyl)-N-(2,3-dichlorophenyl)propionamide
95%
MFCD05156077
Solid
398.28
1g
Videos
SDS