missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(2,3,4-trifluorophenyl)propionamide, 95%

Brand:  Alfa Aesar™ H65121.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15489386

  • £74.73 / 1g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Chemical Identifiers

2-(3-Benzoylphenyl)-N-(2,3,4-trifluorophenyl)propionamide
383.36
MFCD28098751

Specifications

2-(3-Benzoylphenyl)-N-(2,3,4-trifluorophenyl)propionamide
95%
MFCD28098751
Solid
383.36
1g
Videos
SDS