missing translation for 'onlineSavingsMsg'
Learn More

2,3-Benzanthracene, 98%, Thermo Scientific Chemicals

Catalog Number 10468050
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
92-24-0
C18H12
228.29
IFLREYGFSNHWGE-UHFFFAOYSA-N
naphthacene, 2,3-benzanthracene, rubene, benz b anthracene, 2,3-benzanthrene, chrysogen, hydrocarbon, unii-qyj5z6712r, ccris 1183
7080
CHEBI:32600
tetracene
C1=CC=C2C=C3C=C4C=CC=CC4=CC3=CC2=C1
This item is not returnable. View return policy
92-24-0
C18H12
228.29
IFLREYGFSNHWGE-UHFFFAOYSA-N
naphthacene, 2,3-benzanthracene, rubene, benz b anthracene, 2,3-benzanthrene, chrysogen, hydrocarbon, unii-qyj5z6712r, ccris 1183
7080
CHEBI:32600
tetracene
C1=CC=C2C=C3C=C4C=CC=CC4=CC3=CC2=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 05,IV,2545
Merck Index 15,6454
Solubility Information Solubility in water: insoluble. Other solubilities: difficulty soluble in most solvents,soluble in hot xylene
Formula Weight 228.29
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Benzanthracene
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Benzanthracene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'