2,3,6-Trifluorobenzoyl chloride, 98%, ACROS Organics™


Overview
£20.52 valid until 31/03/2017
Log in to get the promotional price



Brand: Acros Organics

Manufacturer Part Number: 312760050

5GR 2,3,6-Trifluorobenzoyl chloride, 98%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 189807-20-3 Weight: 0.05500kg Transport: UN number : 3265 Chem class : 1a Pack group : II


Packaging Glass bottle
Quantity 5g

Product Code. 10766043

Quantity Price
1 £ 51.3 / 5g
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2, 3, 6-Trifluorobenzoyl chloride
CAS 189807-20-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H2ClF3O
Linear Formula F3C6HCOCl
Formula Weight 194.54
MDL Number MFCD00061203

Specification

Assay ≥97.5%
Physical Form Liquid
Formula Weight 194.54
Quantity 5g
Color Clear slightly yellow
CAS Min % 97.5
Linear Formula F3C6HCOCl
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
CAS 189807-20-3
Refractive Index 1.4815 to 1.4835 (20°C, 589nm)
Assay Percent Range 98%
MDL Number MFCD00061203
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Molecular Formula C7H2ClF3O
Health Hazard 1 GHS Signal Word: Danger
CAS Max % 100.0
Density 1.48g/mL
Chemical Name or Material 2, 3, 6-Trifluorobenzoyl chloride
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.