missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trimethylphenol, 98+%, ACROS Organics™

Brand:  Acros Organics 140191000

Code : 29

Additional Details : CAS Number : 697-82-5 Weight : 0.15000kg Transport : UN number : 2430 Chem class : 1a Pack group : III


Quantity 100g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10655382

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

2, 3, 5-Trimethylphenol
697-82-5
100.0
98+%
(CH3)3C6H2OH
MFCD00002228
98+%
98.0
≥98.0%
C9H12O
136.19
Isopseudocumenol

Specifications

2, 3, 5-Trimethylphenol
92° to 95°C
98+%
98.0
Authentic
Needle-Like Crystalline Powder
98+%
(CH3)3C6H2OH
MFCD00002228
White to slightly yellow
100g
697-82-5
100.0
Plastic bottle
≥98.0%
C9H12O
136.19
Isopseudocumenol
Videos
SDS