missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trifluorobenzonitrile, 98%, Thermo Scientific™

Catalog Number p-7025010
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
241154-09-6
C7H2F3N
157.095
MFCD00083539
MMPKVFQMDMBXSG-UHFFFAOYSA-N
benzonitrile, 2,3,5-trifluoro, 2,3,5-trifluorobenzenecarbonitrile, pubchem4823, intermediates-zcf02648, ksc552s4h, 2,3,5-trifluoro-benzonitrile, benzonitrile,2,3,5-trifluoro, 2,3,5-trifluoro benzonitrile
2776998
2,3,5-trifluorobenzonitrile
C1=C(C=C(C(=C1F)F)C#N)F
This item is not returnable. View return policy
241154-09-6
C7H2F3N
157.095
MFCD00083539
MMPKVFQMDMBXSG-UHFFFAOYSA-N
benzonitrile, 2,3,5-trifluoro, 2,3,5-trifluorobenzenecarbonitrile, pubchem4823, intermediates-zcf02648, ksc552s4h, 2,3,5-trifluoro-benzonitrile, benzonitrile,2,3,5-trifluoro, 2,3,5-trifluoro benzonitrile
2776998
2,3,5-trifluorobenzonitrile
C1=C(C=C(C(=C1F)F)C#N)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.47
Quantity 1 g
UN Number UN3276
Formula Weight 157.1
Percent Purity 98%
Chemical Name or Material 2,3,5-Trifluorobenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'