missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trichloropyridine, Thermo Scientific™

Catalog Number 10505924
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
16063-70-0
C5H2Cl3N
182.44
MFCD00043007
CNLIIAKAAMFCJG-UHFFFAOYSA-N
pyridine, 2,3,5-trichloro, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, pubchem1127, g00013-watson-int, acmc-1at5w, 2,3,6-trichloro-pyridine, 2, 3, 5-trichloropyridine, ksc179q6d, tpc-i005
27666
2,3,5-trichloropyridine
C1=C(C=NC(=C1Cl)Cl)Cl
This item is not returnable. View return policy
16063-70-0
C5H2Cl3N
182.44
MFCD00043007
CNLIIAKAAMFCJG-UHFFFAOYSA-N
pyridine, 2,3,5-trichloro, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, pubchem1127, g00013-watson-int, acmc-1at5w, 2,3,6-trichloro-pyridine, 2, 3, 5-trichloropyridine, ksc179q6d, tpc-i005
27666
2,3,5-trichloropyridine
C1=C(C=NC(=C1Cl)Cl)Cl
Melting Point 46°C to 50°C
Boiling Point 219°C
Flash Point 113°C
Packaging Glass Bottle
Quantity 250 g
Formula Weight 182.44
Chemical Name or Material 2, 3, 5-Trichloropyridine
Product Identifier
  • 2,3,5-Trichloropyridine
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'