missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3,5-Trichloropyridine, ≥98%

Brand:  Alfa Aesar™ H64173.36

Code : NEW

Additional Details : CAS Number : 16063-70-0 Weight : 0.50000kg

Quantity 500g

 View more versions of this product

Product Code. 15461259

  • £147.00 / 500g


EU Stock 1
Add to basket

Description

Description

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications

Chemical Identifiers

2,3,5-Trichloropyridine
182.43
119384
16063-70-0
MFCD00043007

Specifications

2,3,5-Trichloropyridine
16063-70-0
≥98%
MFCD00043007
219°C (lit.)
Off-white to pale yellow
500g
Solid
182.43
119384
46° to 50°C (lit.)
Videos
SDS