missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trichloropyridine-4-carboxylic acid, 95%, Thermo Scientific™

Catalog Number 15488386
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
406676-18-4
C6H2Cl3NO2
226.437
MFCD11044390
TYMTYHMQJHSRIF-UHFFFAOYSA-N
2,3,5-trichloroisonicotinic acid, 2,3,5-trichloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 2,3,5-trichloro, trichloroisonicotinicacid, acmc-1bmtq, ksc235e5t, 2,3,5-trichloroisonicotinicacid, 2,3,5-trichloro-isonicotinic acid, 4-carboxy-2,3,5-trichloropyridine, 2,3,5-trichloropyridine-4-carboxylicacid
10944163
2,3,5-trichloropyridine-4-carboxylic acid
C1=C(C(=C(C(=N1)Cl)Cl)C(=O)O)Cl
This item is not returnable. View return policy
406676-18-4
C6H2Cl3NO2
226.437
MFCD11044390
TYMTYHMQJHSRIF-UHFFFAOYSA-N
2,3,5-trichloroisonicotinic acid, 2,3,5-trichloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 2,3,5-trichloro, trichloroisonicotinicacid, acmc-1bmtq, ksc235e5t, 2,3,5-trichloroisonicotinicacid, 2,3,5-trichloro-isonicotinic acid, 4-carboxy-2,3,5-trichloropyridine, 2,3,5-trichloropyridine-4-carboxylicacid
10944163
2,3,5-trichloropyridine-4-carboxylic acid
C1=C(C(=C(C(=N1)Cl)Cl)C(=O)O)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Formula Weight 226.44
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2,3,5-Trichloropyridine-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'