missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trichlorobenzaldehyde, 97%, Thermo Scientific™

Catalog Number 10635472
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
56961-75-2
C7H3Cl3O
209.45
MFCD00010860
DJYRZTCLVDKWBL-UHFFFAOYSA-N
92596
2,3,5-trichlorobenzaldehyde
ClC1=CC(Cl)=C(Cl)C(C=O)=C1
This item is not returnable. View return policy
56961-75-2
C7H3Cl3O
209.45
MFCD00010860
DJYRZTCLVDKWBL-UHFFFAOYSA-N
92596
2,3,5-trichlorobenzaldehyde
ClC1=CC(Cl)=C(Cl)C(C=O)=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 72°C to 76°C
Color White or Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 2.5 g
Beilstein 07, 180
Synonym benzaldehyde, 2,3,5-trichloro, ksc606a4b, acmc-1b126, benzaldehyde,2,3,5-trichloro, djyrztclvdkwbl-uhfffaoysa, 2,3,5-trichlorobenzaldehyde
Formula Weight 209.46
Percent Purity 97%
Physical Form Chunks and/or Powder
Chemical Name or Material 2, 3, 5-Trichlorobenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2,3,5-Trichlorobenzaldehyde
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'