missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trifluorobenzene-1-sulfonyl chloride, 97%, Maybridge

Brand:  Maybridge TL00605EA

Code : 31

Additional Details : CAS Number : 175278-08-7 Weight : 0.06000kg Transport : UN number : 3265 Chem class : 1a Pack group : III

Quantity 10g
Packaging Amber glass bottle

 View more versions of this product

Product Code. 10732513

  • £39.00 / 10g


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Specifications

Chemical Identifiers

2, 3, 4-Trifluorobenzene-1-sulfonyl chloride
97.0
97%
F3C6H2SO2Cl
MFCD00102573
175278-08-7
97%
C6H2ClF3O2S
230.59

Specifications

2, 3, 4-Trifluorobenzene-1-sulfonyl chloride
10g
97.0
97%
C6H2ClF3O2S
230.59
1.6400g/mL
175278-08-7
Amber glass bottle
97%
F3C6H2SO2Cl
MFCD00102573
Videos
SDS