missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3,4-Trifluorobenzamide, 99%

Brand:  Alfa Aesar™ L11304.03

Code : NEW

Additional Details : CAS Number : 207919-09-3 Weight : 0.00100kg

Quantity 1g

Product Code. 15428939

  • £35.72 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Specifications

2,3,4-Trifluorobenzamide
1g
99%
175.11
127° to 129°C
207919-09-3
C7H4F3NO
MFCD00061265
Videos
SDS