missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trifluorobenzaldehyde, 98%, ACROS Organics™

Brand:  Acros Organics 312710010

Code : 29

Additional Details : CAS Number : 161793-17-5 Weight : 0.05100kg Transport : UN number : 1992 Chem class : 3 Pack group : III

Packaging Glass bottle
Quantity 1g

 View more versions of this product

Product Code. 10796233

  • £15.70 / 1g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

2, 3, 4-Trifluorobenzaldehyde
97.5
≥97.5%
C7H3F3O
160.09
161793-17-5
100.0
98%
F3C6H2CHO
MFCD00061230

Specifications

2, 3, 4-Trifluorobenzaldehyde
1g
97.5
Authentic
1.4800 to 1.4820 (20°C, 589nm)
≥97.5%
C7H3F3O
160.09
Clear colorless
161793-17-5
100.0
Glass bottle
Liquid
98%
F3C6H2CHO
MFCD00061230
Videos
SDS