missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3,4-Trifluoro-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide, 95%

Brand:  Alfa Aesar™ H65443.MD

Code : NEW

Additional Details : Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15439646

  • £19.93 / 250mg


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

2,3,4-Trifluoro-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide
305.3
MFCD27975764
N-(5,6,7,8-Tetrahydro-1-naphthyl)-2,3,4-trifluorobenzamide

Specifications

2,3,4-Trifluoro-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide
Solid
305.3
N-(5,6,7,8-Tetrahydro-1-naphthyl)-2,3,4-trifluorobenzamide
250mg
95%
MFCD27975764
Videos
SDS