Alfa Aesar™ 2-(3,4-Difluorophenyl)-5-nitrobenzimidazole, 95%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H64916.03

UNSPSC: 12352101

Code: NEW

Additional Details:
Additional Details: CAS Number: 99585-47-4 Weight: 0.00100kg


Quantity 1g

Product Code. 15412339

Quantity Price
1 £ 119.78 / 1g
Estimated Shipment date
from Supplier 10-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2-(3,4-Difluorophenyl)-5-nitrobenzimidazole
CAS 99585-47-4
Formula Weight 275.21
MDL Number MFCD27665323

Specification

Formula Weight 275.21
Physical Form Solid
Quantity 1g
Assay 95%
MDL Number MFCD27665323
Chemical Name or Material 2-(3,4-Difluorophenyl)-5-nitrobenzimidazole
CAS 99585-47-4
Documentation

Technical documents