missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3,4-Difluorophenyl)-5-methylbenzimidazole, 95%

Brand:  Alfa Aesar™ H64715.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15442029

  • £498.20 / 5g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(3,4-Difluorophenyl)-5-methylbenzimidazole
244.24
MFCD20046535

Specifications

2-(3,4-Difluorophenyl)-5-methylbenzimidazole
Solid
244.24
5g
95%
MFCD20046535
Videos
SDS