missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrahydro-1H-benzo[d]azepine, 97%, Maybridge

Brand:  Maybridge MO08607DE

Code : 31

Additional Details : CAS Number : 4424-20-8 Weight : 0.05500kg Transport : UN number : 2735 Chem class : 1b Pack group : III

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10096914

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

2,3,4,5-Tetrahydro-1H-benzo[d]azepine
97%
147.22
97%
C10H13N

Specifications

2,3,4,5-Tetrahydro-1H-benzo[d]azepine
4424-20-8
97%
C10H13N
5g
Amber Glass Bottle
97%
147.22
Videos
SDS