missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrafluorobenzamide, 97+%, Thermo Scientific™

Catalog Number p-7026760
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
16582-94-8
C7H3F4NO
193.10
MFCD01631457
XMFMIKSWEJRJQE-UHFFFAOYSA-N
acmc-1c7cs, benzamide,2,3,4,5-tetrafluoro
2776581
2,3,4,5-tetrafluorobenzamide
NC(=O)C1=CC(F)=C(F)C(F)=C1F
This item is not returnable. View return policy
16582-94-8
C7H3F4NO
193.10
MFCD01631457
XMFMIKSWEJRJQE-UHFFFAOYSA-N
acmc-1c7cs, benzamide,2,3,4,5-tetrafluoro
2776581
2,3,4,5-tetrafluorobenzamide
NC(=O)C1=CC(F)=C(F)C(F)=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 132°C to 134°C
Quantity 1 g
Formula Weight 193.1
Percent Purity ≥97%
Chemical Name or Material 2,3,4,5-Tetrafluorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'