missing translation for 'onlineSavingsMsg'
Learn More

2,3:4,5-Di-O-isopropylidene-D-arabitol, 98%, Thermo Scientific Chemicals

Catalog Number p-7043254
Click to view available options
Quantity:
500 mg
1 g
This item is not returnable. View return policy
19139-74-3
C11H20O5
232.28
MFCD00063387
FBIWGPWXTDOFQR-UHFFFAOYNA-N
2-o,3-o:4-o,5-o-diisopropylidene-d-arabinitol
11042611
[(4R,5R)-5-[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
CC1(C)OCC(O1)C1OC(C)(C)OC1CO
This item is not returnable. View return policy
19139-74-3
C11H20O5
232.28
MFCD00063387
FBIWGPWXTDOFQR-UHFFFAOYNA-N
2-o,3-o:4-o,5-o-diisopropylidene-d-arabinitol
11042611
[(4R,5R)-5-[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
CC1(C)OCC(O1)C1OC(C)(C)OC1CO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Protect from heat. Incompatible with heat and oxidizing agents.
TRUSTED_SUSTAINABILITY
Color Colorless
Quantity 500 mg
Formula Weight 232.27
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material 2,3:4,5-Di-O-isopropylidene-D-arabitol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'