missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorophenylacetonitrile, 98%, Thermo Scientific Chemicals

Catalog Number p-7026306
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
653-30-5
C8H2F5N
207.103
MFCD00013814
YDNOJUAQBFXZCR-UHFFFAOYSA-N
2,3,4,5,6-pentafluorophenylacetonitrile, pentafluorophenylacetonitrile, pentafluorophenyl acetonitrile, 2-2,3,4,5,6-pentafluorophenyl acetonitrile, 2,3,4,5,6-pentafluorobenzyl cyanide, 2-perfluorophenyl acetonitrile, benzeneacetonitrile, 2,3,4,5,6-pentafluoro, 2-2,3,4,5,6-pentafluorophenyl ethanenitrile, acmc-1ay9x, 2-pentafluorophenyl acetonitrile
69554
2-(2,3,4,5,6-pentafluorophenyl)acetonitrile
C(C#N)C1=C(C(=C(C(=C1F)F)F)F)F
This item is not returnable. View return policy
653-30-5
C8H2F5N
207.103
MFCD00013814
YDNOJUAQBFXZCR-UHFFFAOYSA-N
2,3,4,5,6-pentafluorophenylacetonitrile, pentafluorophenylacetonitrile, pentafluorophenyl acetonitrile, 2-2,3,4,5,6-pentafluorophenyl acetonitrile, 2,3,4,5,6-pentafluorobenzyl cyanide, 2-perfluorophenyl acetonitrile, benzeneacetonitrile, 2,3,4,5,6-pentafluoro, 2-2,3,4,5,6-pentafluorophenyl ethanenitrile, acmc-1ay9x, 2-pentafluorophenyl acetonitrile
69554
2-(2,3,4,5,6-pentafluorophenyl)acetonitrile
C(C#N)C1=C(C(=C(C(=C1F)F)F)F)F

2,3,4,5,6-Pentafluorophenylacetonitrile is used as medicine and liquid crystal materials intermediates. Ortho or para substituted dimers and a mixture of trimers, tetramers and pentamers of PFPA coulb be obtained from 2,3,4,5,6-Pentafluorophenylacetonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5,6-Pentafluorophenylacetonitrile is used as medicine and liquid crystal materials intermediates. Ortho or para substituted dimers and a mixture of trimers, tetramers and pentamers of PFPA coulb be obtained from 2,3,4,5,6-Pentafluorophenylacetonitrile.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.61
Boiling Point 110°C to 111°C (17 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.439
Quantity 1 g
UN Number UN3276
Beilstein 1882341
Solubility Information Insoluble in water.
Formula Weight 207.1
Percent Purity 98%
Chemical Name or Material 2,3,4,5,6-Pentafluorophenylacetonitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'