missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluoroaniline, 97%, Thermo Scientific Chemicals

Catalog Number p-3779384
Click to view available options
Quantity:
5 g
10 g
50 g
250 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
771-60-8
C6H2F5N
183.08
MFCD00007643
NOXLGCOSAFGMDV-UHFFFAOYSA-N
pentafluoroaniline, pentafluorophenylamine, aminopentafluorobenzene, benzenamine, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluorobenzenamine, aniline, 2,3,4,5,6-pentafluoro, unii-ms06il3y2q, ms06il3y2q, 2,3,4,5,6-pentafluorophenylamine, pubchem1546
13040
2,3,4,5,6-pentafluoroaniline
NC1=C(F)C(F)=C(F)C(F)=C1F
This item is not returnable. View return policy
771-60-8
C6H2F5N
183.08
MFCD00007643
NOXLGCOSAFGMDV-UHFFFAOYSA-N
pentafluoroaniline, pentafluorophenylamine, aminopentafluorobenzene, benzenamine, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluorobenzenamine, aniline, 2,3,4,5,6-pentafluoro, unii-ms06il3y2q, ms06il3y2q, 2,3,4,5,6-pentafluorophenylamine, pubchem1546
13040
2,3,4,5,6-pentafluoroaniline
NC1=C(F)C(F)=C(F)C(F)=C1F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 33°C to 38°C
Color Salmon to White
Density 73g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula C6F5NH2
Quantity 5 g
Formula Weight 183.08
Percent Purity ≥96.0%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 2,3,4,5,6-Pentafluoroaniline
Show More Show Less
compliance-icons
Product Identifier
  • 2,3,4,5,6-Pentafluoroaniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'