missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-[2-(Trifluoromethyl)phenyl]ethylamine, 95%

Brand:  Alfa Aesar™ H64492.06

Code : NEW

Additional Details : CAS Number : 774-18-5 Weight : 0.00500kg Transport : UN number : 2735 Chem class : 1b Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15451729

  • £60.10 / 5g


Estimated Shipment date
from Supplier 19-03-2018
Add to basket

Specifications

Chemical Identifiers

2-[2-(Trifluoromethyl)phenyl]ethylamine
189.18
UN2735
2-(Trifluoromethyl)phenethylamine
774-18-5
MFCD00128232
7202211

Specifications

2-[2-(Trifluoromethyl)phenyl]ethylamine
Pale yellow
Liquid
189.18
UN2735
2-(Trifluoromethyl)phenethylamine
774-18-5
5g
95%
MFCD00128232
7202211
Videos
SDS