missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Thiodiethanethiol, tech. 85%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
2,2?-Thiodiethanol is used in the synthesis of novel luminescent polynuclear gold(I) phosphine complexes 2,2'-Thiodiethanethiol is used for extraction photometric determination of Ni, gives color reaction with metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2′-Thiodiethanol is used in the synthesis of novel luminescent polynuclear gold(I) phosphine complexes 2,2′-Thiodiethanethiol is used for extraction photometric determination of Ni, gives color reaction with metals.
Solubility
Difficult to mix.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2,2′-Thiodiethanol is used in the synthesis of novel luminescent polynuclear gold(I) phosphine complexes 2,2′-Thiodiethanethiol is used for extraction photometric determination of Ni, gives color reaction with metals.
Solubility
Difficult to mix.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Density | 1.183 |
Boiling Point | 135°C to 136°C (10 mmHg) |
Flash Point | 90°C (194°F) |
Refractive Index | 1.5955 |
Quantity | 10 g |
Solubility Information | Difficult to mix. |
IUPAC Name | 2-(2-sulfanylethylsulfanyl)ethanethiol |
Formula Weight | 154.32 |
Percent Purity | ≥90% |
Chemical Name or Material | 2,2'-Thiodiethanethiol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'