missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Thienylsulfonyl)ethanethioamide, 97%, Thermo Scientific™

Catalog Number p-2696113
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
175202-34-3
C6H7NO2S3
221.307
MFCD00052277
YNPIRUTXXLFXRU-UHFFFAOYSA-N
2-2-thienylsulfonyl ethanethioamide, 2-thiophene-2-sulfonyl ethanethioamide, ethanethioamide,2-2-thienylsulfonyl, maybridge1_004617, 2-2-thienyl sulfonyl thioacetamide, 2-thiophen-2-ylsulfonyl ethanethioamide, 2-thien-2-yl sulphonyl ethanethioamide, 2-2-amino-2-thioxoethyl sulphonyl thiophene, 2-amino-1-2-thienylsulfonyl ethane-2-thione
2778709
2-thiophen-2-ylsulfonylethanethioamide
C1=CSC(=C1)S(=O)(=O)CC(=S)N
This item is not returnable. View return policy
175202-34-3
C6H7NO2S3
221.307
MFCD00052277
YNPIRUTXXLFXRU-UHFFFAOYSA-N
2-2-thienylsulfonyl ethanethioamide, 2-thiophene-2-sulfonyl ethanethioamide, ethanethioamide,2-2-thienylsulfonyl, maybridge1_004617, 2-2-thienyl sulfonyl thioacetamide, 2-thiophen-2-ylsulfonyl ethanethioamide, 2-thien-2-yl sulphonyl ethanethioamide, 2-2-amino-2-thioxoethyl sulphonyl thiophene, 2-amino-1-2-thienylsulfonyl ethane-2-thione
2778709
2-thiophen-2-ylsulfonylethanethioamide
C1=CSC(=C1)S(=O)(=O)CC(=S)N
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 221.32
Percent Purity 97%
Chemical Name or Material 2-(2-Thienylsulfonyl)ethanethioamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'