missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(2-Thienyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62735.06

Code : NEW

Additional Details : CAS Number : 32451-19-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15439218

  • £543.78 / 5g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

2-(2-Thienyl)thiazolidine-4-carboxylic acid
32451-19-7
215.29
MFCD00022489

Specifications

2-(2-Thienyl)thiazolidine-4-carboxylic acid
32451-19-7
97%
MFCD00022489
5g
Solid
215.29
Videos
SDS