missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Thienyl)ethyl isocyanate, 95+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Synonym | 2-2-isocyanatoethyl thiophene, 2-2-thienyl ethyl isocyanate, 2-thien-2-yl ethyl isocyanate, thiophene,2-2-isocyanatoethyl, 2-2-isocyanato-ethyl-thiophene, 2-2-thienyl ethanisocyanate, 2-2-isocyanatoethyl-thiophene, thiophene, 2-2-isocyanatoethyl, 2-thien-2-yl ethylisocyanate |
Formula Weight | 153.2 |
Percent Purity | 95% |
Chemical Name or Material | 2-(2-Thienyl)ethyl isocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'