missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Thienyl)ethanol, 98%, Thermo Scientific Chemicals

Catalog Number p-2689610
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5402-55-1
C6H8OS
128.19
MFCD00005462
VMJOFTHFJMLIKL-UHFFFAOYSA-N
2-thiopheneethanol, 2-2-thienyl ethanol, thiophene-2-ethanol, 2-thiophen-2-yl ethanol, 2-2-hydroxyethyl thiophene, 2-thiophen-2-yl ethan-1-ol, ethanol, 2-2-thienyl, 2-thiophenethanol, 2-2-thienyl-ethanol, 2-thienylethanol
79400
2-thiophen-2-ylethanol
OCCC1=CC=CS1
This item is not returnable. View return policy
5402-55-1
C6H8OS
128.19
MFCD00005462
VMJOFTHFJMLIKL-UHFFFAOYSA-N
2-thiopheneethanol, 2-2-thienyl ethanol, thiophene-2-ethanol, 2-thiophen-2-yl ethanol, 2-2-hydroxyethyl thiophene, 2-thiophen-2-yl ethan-1-ol, ethanol, 2-2-thienyl, 2-thiophenethanol, 2-2-thienyl-ethanol, 2-thienylethanol
79400
2-thiophen-2-ylethanol
OCCC1=CC=CS1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Brown or Gray-Green to Yellow
Density 1.1530g/mL
Boiling Point 108°C (13.0 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5490 to 1.552
Quantity 25 g
Beilstein 17, V,3, 379
Specific Gravity 1.153
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in mek,alcohol,organic halogen solvents
Formula Weight 128.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(2-Thienyl)ethanol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-(2-Thienyl)ethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'