missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(2-Pyridyl)thiazolidine, 97%

Brand:  Alfa Aesar™ H62437.03

Code : NEW

Additional Details : CAS Number : 700-94-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15488938

  • £109.18 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(2-Pyridyl)thiazolidine
166.24
2-(2-Thiazolidinyl)pyridine
700-94-7
MFCD01691462

Specifications

2-(2-Pyridyl)thiazolidine
1g
Liquid
166.24
2-(2-Thiazolidinyl)pyridine
Dark red
700-94-7
97%
MFCD01691462
Videos
SDS