missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(2-Pyridyl)thiazolidine, 97%

Brand:  Alfa Aesar™ H62437.06

Code : NEW

Additional Details : CAS Number : 700-94-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15498938

  • £543.78 / 5g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

2-(2-Pyridyl)thiazolidine
166.24
2-(2-Thiazolidinyl)pyridine
700-94-7
MFCD01691462

Specifications

2-(2-Pyridyl)thiazolidine
5g
Liquid
166.24
2-(2-Thiazolidinyl)pyridine
Dark red
700-94-7
97%
MFCD01691462
Videos
SDS