Learn More
Alfa Aesar™ 2-(2-Pyridyl)thiazolidine-4-carboxylic acid, 97%
Brand: Alfa Aesar™ H62636.06
Code : NEW
Additional Details : CAS Number : 84682-22-4 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
2-(2-Pyridyl)thiazolidine-4-carboxylic acid | |
84682-22-4 | |
210.25 | |
MFCD09949861 |
Specifications
2-(2-Pyridyl)thiazolidine-4-carboxylic acid | |
84682-22-4 | |
97% | |
MFCD09949861 |
5g | |
Solid | |
210.25 |