missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Propynyloxy)tetrahydro-2H-pyran, 98%, Thermo Scientific Chemicals

Catalog Number p-3779515
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6089-04-9
C8H12O2
140.18
MFCD00006604
HQAXHIGPGBPPFU-UHFFFAOYNA-N
tetrahydro-2-2-propynyloxy-2h-pyran, 2-2-propynyloxy tetrahydropyran, 2-propargyloxane, 2-prop-2-yn-1-yloxy tetrahydro-2h-pyran, 2-propargyloxy tetrahydropyran, propargyl 2-tetrahydropyranyl ether, propargyl alcohol tetrahydropyranyl ether, 2h-pyran, tetrahydro-2-2-propynyloxy, 2-propynyl 2-pyranyl ether, 3-2'-tetrahydropyranyloxy propyne
98609
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6089-04-9
C8H12O2
140.18
MFCD00006604
HQAXHIGPGBPPFU-UHFFFAOYNA-N
tetrahydro-2-2-propynyloxy-2h-pyran, 2-2-propynyloxy tetrahydropyran, 2-propargyloxane, 2-prop-2-yn-1-yloxy tetrahydro-2h-pyran, 2-propargyloxy tetrahydropyran, propargyl 2-tetrahydropyranyl ether, propargyl alcohol tetrahydropyranyl ether, 2h-pyran, tetrahydro-2-2-propynyloxy, 2-propynyl 2-pyranyl ether, 3-2'-tetrahydropyranyloxy propyne
98609

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9970g/mL
Boiling Point 63°C to 65°C (9.0 mmHg)
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4560 to 1.458
Quantity 5 g
Specific Gravity 0.997
Formula Weight 140.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(2-Propynyloxy)tetrahydro-2H-pyran
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-(2-Propynyloxy)- tetrahydro-2H-pyran
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'