missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(2-Phenylethyl)thiazolidine, 97%

Brand:  Alfa Aesar™ H62323.06

Code : NEW

Additional Details : CAS Number : 55089-15-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15438858

  • £460.04 / 5g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

2-(2-Phenylethyl)thiazolidine
193.31
2-Phenethylthiazolidine
55089-15-1
MFCD01681080

Specifications

2-(2-Phenylethyl)thiazolidine
55089-15-1
97%
MFCD01681080
5g
Liquid
193.31
2-Phenethylthiazolidine
Videos
SDS